Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOXD4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548772

Catalog No. PA548772

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with chicken and mouse based on sequence homology.

Sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis.
Specifications

Specifications

HOXD4
Polyclonal
PBS with 0.09% sodium azide
P09016
Homeobox protein Hox-D4, Homeobox protein HHOC13, Homeobox protein Hox-4B, Homeobox protein Hox-51, HOXD4, HOX4B
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3233
Western Blot
Unconjugated
HOXD4
Liquid
HOXD4
KLH conjugated synthetic peptide between 195-223 amino acids from the C-terminal region of human HOXD4.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit