Learn More
HOXD8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 185881AP
Catalog No. 50-173-2182
Add to cart
Description
Description
This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, located on different chromosomes, consisting of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXD genes located in a cluster on chromosome 2. Deletions that remove the entire HOXD gene cluster or the 5 ft. end of this cluster have been associated with severe limb and genital abnormalities. In addition to effects during embryogenesis, this particular gene may also play a role in adult urogenital tract function.
Specifications
Specifications
HOXD8
0.26 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P13378, P23463
homeobox D8, Homeobox protein Hox 4E, Homeobox protein Hox 5.4, Homeobox protein Hox D8, HOX4, HOX4E, HOX5.4, HOXD8
Rabbit
IgG
150 μL
-20°C
Primary
100912936, 15437, 3234
Western Blot
Unconjugated
HOXD8
Liquid
HOXD8
HOXD8 Fusion Protein Ag13245
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat