Learn More
Aviva Systems Biology Corp HP1BP3 Antibody - middle region (ARP56875_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP56875_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HP1BP3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HP1BP3. Alternate names: HP1-BP74, MGC43701, RP5-930J4.3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62023