Learn More
HP1BP3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 245561AP
Catalog No. 50-173-4419
Add to cart
Description
Description
HP1BP3 is the component of heterochromatin, may be involved in chromatin structure and function.
Specifications
Specifications
HP1BP3
0.42 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q3TEA8, Q5SSJ5
HP1 BP74, HP1BP3, Protein HP1 BP74
Rabbit
IgG
150 μL
-20°C
Primary
15441, 50809
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
HP1BP3
Liquid
HP1BP3
HP1BP3 Fusion Protein Ag18709
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse