Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HPa2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576358
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
This gene encodes a heparanase enzyme. The encoded protein is a endoglycosidase that degrades heparin sulfate proteoglycans located on the extracellular matrix and cell surface. This protein may be involved in biological processes involving remodeling of the extracellular matrix including angiogenesis and tumor progression. Alternate splicing results in multiple transcript variants.
Specifications
HPa2 | |
Polyclonal | |
Unconjugated | |
HPSE2 | |
Gm968; heparanase 2; heparanase 2 (inactive); heparanase 3; heparanase-2; heparanase-like protein; Hpa2; HPR2; Hpse2; Inactive heparanase-2; UFS; UFS1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
368128, 545291, 60495 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
B2RY83, Q8WWQ2 | |
HPSE2 | |
Recombinant protein of human HPSE2. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction