Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HPCAL4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113513
Description
HPCAL4 Polyclonal Antibody for Western Blot, ELISA
May be involved in the calcium-dependent regulation of rhodopsin phosphorylation.
Specifications
HPCAL4 | |
Polyclonal | |
Unconjugated | |
HPCAL4 | |
AI846570; hippocalcin like 4; hippocalcin-like 4; hippocalcin-like protein 4; HLP4; HPCAL4; Neural visinin-like protein 2; NVP-2 | |
Rabbit | |
Protein G | |
RUO | |
170638, 51440 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q8BGZ1, Q9UM19 | |
HPCAL4 | |
Recombinant Human Hippocalcin-like protein 4 protein (2-191AA). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction