Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPd1 Mouse anti-Human, PE/Atto594, Clone: DHIC2-4A10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP118953PEATT594

Catalog No. N118953P594


Add to cart

Description

Description

HPd1 Monoclonal antibody specifically detects HPd1 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

HPd1
DHIC2-4A10
Flow Cytometry
Conjugated, Supernatant
Mouse
IgG1
0.1 mL
Primary
Specific for duct cell types.
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
DHIC2-4A10, HPd1, human pancreatic duct cell
Human pancreatic enriched duct cells
Tissue culture supernatant
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit