Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPd2 Mouse anti-Human, Allophycocyanin/Cy7, Clone: DHIC2-5H10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP118955APCCY7

Catalog No. N11895APCY7

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

HPd2 Monoclonal antibody specifically detects HPd2 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

HPd2
DHIC2-5H10
Flow Cytometry
Conjugated, Supernatant
Mouse
IgM
0.1 mL
Primary
Specific for duct cell types.
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
human pancreatic duct cell
Human pancreatic enriched duct cells
Tissue culture supernatant
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit