Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPRG Mouse anti-Human, Alexa Fluor™ 594, Clone: MM0331-6Z48, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP212359AF594

Catalog No. NP21239A94


Add to cart

Description

Description

HPRG Monoclonal antibody specifically detects HPRG in Human samples. It is validated for Western Blot.
Specifications

Specifications

HPRG
MM0331-6Z48
Western Blot
Conjugated, Purified
HRG
Human HRG
Protein G purified
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
DKFZp779H1622, Histidine-proline-rich glycoprotein, histidine-rich glycoprotein, HPRGHRGP
Mouse
IgG1
0.1 mL
Primary
3273.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit