Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HPRG Rat anti-Mouse, DyLight 550, Clone: RM0091-9G42, Novus Biologicals
Rat Monoclonal Antibody
Manufacturer: Novus Biologicals NBP122407R
Description
HPRG Monoclonal antibody specifically detects HPRG in Mouse samples. It is validated for Western Blot.Specifications
HPRG | |
RM0091-9G42 | |
Western Blot | |
Q9ESB3 | |
DKFZp779H1622, Histidine-proline-rich glycoprotein, histidine-rich glycoprotein, HPRGHRGP | |
Rat | |
IgG2 | |
0.1 ml | |
Primary | |
3273 | |
Mouse |
Western Blot | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Purified | |
HRG | |
Mouse recombinant protein of HPRG | |
Protein G purified | |
Store at 4C in the dark. | |
Monoclonal | |
This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein of HPRG. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title