Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HPRT Rabbit anti-Human, Mouse, Rat, Zebrafish, Clone: JU03-26, Novus Biologicals
Rabbit Monoclonal Antibody
Manufacturer: Novus Biologicals NBP275528
Description
HPRT Monoclonal antibody specifically detects HPRT in Human, Mouse, Rat, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin).Specifications
HPRT | |
JU03-26 | |
Unconjugated | |
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.05% Sodium Azide | |
EC 2.4.2.8, HGPRTHPRTHGPRTase, hypoxanthine phosphoribosyltransferase 1, hypoxanthine-guanine phosphoribosyltransferase | |
Rabbit | |
IgG | |
Protein A purified | |
Lipid and Metabolism | |
Primary | |
3251.0 |
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1 mg/mL | |
Western Blot 1:500-1:2000 Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:10-1:50 Immunohistochemistry-Paraffin 1:50-1:200 | |
Purified | |
HPRT1 | |
Recombinant Protein | |
25 kDa | |
100 μL | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
Monoclonal | |
Human, Mouse, Rat, Zebrafish |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title