Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPRT Mouse anti-Human, Janelia Fluor™ 549, Clone: OTI2F7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270934JF549

Catalog No. N270934J549


Add to cart

Description

Description

HPRT Monoclonal antibody specifically detects HPRT in Human samples. It is validated for Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

HPRT
OTI2F7
Immunocytochemistry/Immunofluorescence
Affinity Purified, Conjugated
HPRT1
Full length recombinant protein of human HPRT (NP_000185) produced in HEK293T cells.
Immunogen affinity purified
Lipid and Metabolism
Primary
3251.0
Immunocytochemistry, Immunofluorescence
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
EC 2.4.2.8, HGPRTHPRTHGPRTase, hypoxanthine phosphoribosyltransferase 1, hypoxanthine-guanine phosphoribosyltransferase
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit