Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPRT1 Mouse anti-Human, Clone: OTI2D5, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526295

Catalog No. PIMA526295


Add to cart

Description

Description

The HPRT1 gene provides instructions for making an enzyme called hypoxanthine phosphoribosyltransferase 1. This enzyme allows cells to recycle purines, some of the building blocks of DNA and its chemical cousin RNA.The enzyme hypoxanthine-guanine phosphoribosyltrasferase (E.C.2.4.2.8., HPRT) plays a crucial role in uric acid synthesis and purine metabolism.This enzyme catalyzes the conversion of hypoxanthine and guanine to inosine monophosphate (IMP) and guanosine monophosphate (GMP), respectively, and uses phosphoribosylpyrophosphate (PRPP) as a cosubstrate and as a source of energy. This pathway is also known as the purine salvage pathway because it allows cells to reuse purine compounds to build DNA and RNA.
Specifications

Specifications

HPRT1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P00492
HGPRT;HGPRTase;HPRT 1;HPRT_HUMAN;HPRT1;Hypoxanthine guanine phosphoribosyltransferase;Hypoxanthine phosphoribosyltransferase 1 (Lesch Nyhan syndrome);Hypoxanthine phosphoribosyltransferase 1;Hypoxanthine-guanine phosphoribosyltransferase;
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3251
Immunocytochemistry, Immunofluorescence, Western Blot
OTI2D5
Unconjugated
HPRT1
Liquid
HPRT1
Full length human recombit protein of HPRT1 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit