Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPSC152 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen HPSC152
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB093941
View Documents
Novus Biologicals
NBP303251100UL
100 μg
Each for $320.71
Add to cart
 
NB093942
View Documents
Novus Biologicals
NBP30325120UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

HPSC152 Polyclonal antibody specifically detects HPSC152 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

HPSC152
Unconjugated
Rabbit
Stem Cell Markers
PBS with 50% glycerol, pH7.3.
51504
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot
Purified
RUO
Human, Mouse, Rat
HSPC152, HSPC170, TRM112, TRM112-like protein, TRMT11-2, tRNA methyltransferase 112 homolog, tRNA methyltransferase 11-2 homolog (S. cerevisiae)
Recombinant fusion protein containing a sequence corresponding to amino acids 1-125 of human HPSC152 (NP_057488.1).
Polyclonal
Affinity purified
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit