Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HPV-16 Rabbit anti-Human, Unconjugated, Clone: HPV16/2058R, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation RAB00696
Description
HPV-16 Monoclonal antibody specifically detects HPV-16 in Human samples. It is validated for IHC-P.Specifications
HPV-16 | |
Monoclonal | |
Unconjugated | |
Human papilloma virus type 16, major capsid protein L1 | |
Primary | |
Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Immunohistochemistry (Paraffin) | |
HPV16/2058R | |
Rabbit | |
100 μg | |
Human | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction