Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPV Type 16 Mouse anti-Human papilloma virus (HPV), Clone: SPM405, Abnova™

Mouse monoclonal antibody raised against human papilloma virus type 16.

Manufacturer:  Abnova Corporation MAB13200

Catalog No. 89-934-299


Add to cart

Description

Description

Specifications

Specifications

HPV Type 16
SPM405
Mouse monoclonal antibody raised against human papilloma virus type 16.
In 10mM PBS.
Mouse
IgG2a, kappa
100 ug
Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
Recombinant major capsid protein L1 from human papilloma virus type 16.
Protein A/G purification
RUO
Primary
Human papilloma virus
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit