Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPV16/HPV18 E6 Mouse anti-Virus, Clone: HPV16/1295 + HPV18/1297, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30767510UG

 View more versions of this product

Catalog No. NB122823


Add to cart

Description

Description

HPV16/HPV18 E6 Monoclonal antibody specifically detects HPV16/HPV18 E6 in Virus samples. It is validated for ELISA
Specifications

Specifications

HPV16/HPV18 E6
HPV16/1295 + HPV18/1297
Unconjugated
10 mM PBS with 0.05% BSA
E6, Human Papilloma Virus, Human papillomavirus type 16 E6, Human papillomavirus type 18 E6, Protein E6
HPV18 E6-beta-galactosidase fusion protein
Protein A or G purified
RUO
Primary
Virus
ELISA
0.2 mg/ml
ELISA
Purified
Mouse
IgG1 Kappa
100 μg
Store at 4°C.
Monoclonal
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit