Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPV16 L1 Mouse anti-Human, Virus, Clone: HPV16/1296, Novus Biologicals™

Mouse Monoclonal Antibody

$152.22 - $378.35

Specifications

Antigen HPV16 L1
Clone HPV16/1296
Host Species Mouse
Isotype IgG2a Kappa
Gene Alias HPV-16, HPV-16 capsid, HPV16 capsid protein, HPV16 L1, HPV16 major capsid protein L1, Human papillomavirus type 16 L1, Human papillomavirus type 16 major capsid protein L1, Major capsid protein, Major capsid protein L1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB25337420 View Documents Novus Biologicals
NBP25337420UG
20 ug Each for $152.22
Add to cart
 
NB253374100 View Documents Novus Biologicals
NBP253374100UG
100 ug Each for $378.35
Add to cart
 
NB253374200 View Documents Novus Biologicals
NBP253374200UG
200 ug This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

HPV16 L1 Monoclonal antibody specifically detects HPV16 L1 in Virus samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

HPV16 L1
Mouse
HPV-16, HPV-16 capsid, HPV16 capsid protein, HPV16 L1, HPV16 major capsid protein L1, Human papillomavirus type 16 L1, Human papillomavirus type 16 major capsid protein L1, Major capsid protein, Major capsid protein L1
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Store at 4C.
Monoclonal
0.2 mg/mL
HPV16/1296
IgG2a Kappa
Human, Virus
RUO
Purified
Primary
10mM PBS with 0.05% BSA with 0.05% Sodium Azide

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit