Learn More
HPV16 L1 Mouse anti-Virus, Alexa Fluor™ 594, Clone: HPV16/1296, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP254510AF594
Catalog No. NP2410A94
Add to cart
Description
Description
HPV16 L1 Monoclonal antibody specifically detects HPV16 L1 in Virus samples. It is validated for Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin).
Specifications
Specifications
HPV16 L1
HPV16/1296
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
HPV-16, HPV-16 capsid, HPV16 capsid protein, HPV16 L1, HPV16 major capsid protein L1, Human papillomavirus type 16 L1, Human papillomavirus type 16 major capsid protein L1, Major capsid protein, Major capsid protein L1
Human papilloma virus type 16
0.1 mL
Primary
Virus
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
Mouse
IgG2a Kappa
Store at 4°C in the dark.
Monoclonal