Learn More
HPV16 Mouse anti-Virus, Alexa Fluor 700, Clone: 716-325, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NB11017341AF700
Catalog No. NB11017341N
Add to cart
Description
Description
HPV16 E7 Monoclonal specifically detects HPV16 E7 in Virus samples. It is validated for Western Blot, ELISA, Sandwich ELISA.
Specifications
Specifications
HPV16
716-325
Western Blot, ELISA, Sandwich ELISA
Purified
Mouse
IgG2a
0.2 ml
Store at 4C in the dark.
Monoclonal
Virus
Western Blot, ELISA
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
Human Papilloma Virus Type 16
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with HPV oncoprotein E7, type 16.
Protein G purified
RUO
Primary
HPV oncoprotein E7, types 16 & 18 (both monomer and dimer forms according WB)