Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPV16 Mouse anti-Virus, Clone: HPV16/1295, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30784410UG

 View more versions of this product

Catalog No. NB123161



Description

Description

HPV16 Monoclonal specifically detects HPV16 in Virus samples. It is validated for Immunohistochemistry-Paraffin.
Specifications

Specifications

HPV16
HPV16/1295
Unconjugated
10 mM PBS with 0.05% BSA
Human Papilloma Virus Type 16
HPV-16 E6 peptide (MHQKRTAMFQDPPQERPRKLPQLC)
Protein A or G purified
RUO
Primary
Virus
Immunohistochemistry (Paraffin)
0.2 mg/ml
Immunohistochemistry-Paraffin 1-2 ug/ml
Purified
Mouse
IgG1 Kappa
100 μg
Store at 4°C.
Monoclonal
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit