Learn More
HPV16 Mouse anti-Virus, Clone: SPM257, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP30779110UG
 View more versions of this product
Catalog No. NB123055
Description
Description
HPV16 Monoclonal antibody specifically detects HPV16 in Virus samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
HPV16
SPM257
Unconjugated
10 mM PBS with 0.05% BSA
Human Papilloma Virus Type 16
Human HPV16, major capsid protein L1
Protein A or G purified
RUO
Primary
Virus
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
0.2 mg/ml
Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin
Purified
Mouse
IgG2a Kappa
100 μg
Store at 4°C.
Monoclonal