Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HPV16 Rabbit anti-Virus, Clone: HPV16/2058R, Novus Biologicals™
Rabbit Monoclonal Antibody
$301.00
Specifications
Antigen | HPV16 |
---|---|
Clone | HPV16/2058R |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
Applications | Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB122094
|
Novus Biologicals
NBP30731020UG |
20 μg |
Each of 1 for $301.00
|
|
|||||
Description
HPV16 Monoclonal specifically detects HPV16 in Virus samples. It is validated for Immunohistochemistry-Paraffin.Specifications
HPV16 | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Virus | |
Human Papilloma Virus Type 16 | |
IgG | |
Protein A or G purified |
HPV16/2058R | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
Human HPV16, major capsid protein L1 | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title