Learn More
HPV16E1+E4 Mouse anti-Virus, Janelia Fluor 646, Clone: TVG 401, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP250426JF646
Catalog No. NBP50426646
Add to cart
Description
Description
HPV16E1+E4 Monoclonal antibody specifically detects HPV16E1+E4 in Virus samples. It is validated for Immunohistochemistry.
Specifications
Specifications
HPV16E1+E4
TVG 401
Immunohistochemistry
Purified
Mouse
IgG1
0.1mL
Primary
Virus
Immunohistochemistry
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
ATP dependent helicase E1, E1, E4, HPV 16 ATP dependent helicase E1, HPV 16 E1, HPV 16 E4, HPV 16 replication protein E1, HPV16 ATP dependent helicase E1, HPV16 E1, HPV16 E4, HPV16 replication protein E1, Human papilloma virus type 16, Human papilloma virus type 16 ATP dependent helicase E1, Human papilloma virus type 16 E1, Human papilloma virus type 16 E4, Human papilloma virus type 16 replication protein E1, Human papillomavirus type 16, Human papillomavirus type 16 ATP dependent helicase E1, Human papillomavirus type 16 E1, Human papillomavirus type 16 E4, Human papillomavirus type 16 replication protein E1, Replication protein, Replication protein E1
Antigen for hybridoma production was expressed as a b galactosidase fusion protein using the pEX expression system and was consequently cleaved to release the E1/\E4 polypeptide.
Protein G purified
Store at 4°C in the dark.
Monoclonal