Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPV52 L1 Mouse anti-Virus, Clone: 8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP31137625UL

 View more versions of this product

Catalog No. NB121461


Add to cart

Description

Description

HPV52 L1 Monoclonal antibody specifically detects HPV52 L1 in Virus samples. It is validated for ELISA
Specifications

Specifications

HPV52 L1
8
ELISA -1:1000-1:2000
Purified
Recombinant Human Papilloma Virus type 52 L1 virus like particle (HPV52 L1 VLP)
Protein A purified
RUO
Primary
Virus
ELISA
Unconjugated
PBS, pH7.0
Mouse
IgG2b
25 μg
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit