Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPV6 L1 Rabbit anti-Virus, Clone: 1, Novus Biologicals™

Rabbit Monoclonal Antibody

$192.00 - $460.15

Specifications

Antigen HPV6 L1
Clone 1
Dilution ELISA -1:5000-1:10000
Applications ELISA
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB121466
View Documents
Novus Biologicals
NBP311379100UL
100 μg
Each for $460.15
Add to cart
 
NB121467
View Documents
Novus Biologicals
NBP31137925UL
25 μg
Each for $192.00
Add to cart
 
Description

Description

HPV6 L1 Monoclonal antibody specifically detects HPV6 L1 in Virus samples. It is validated for ELISA
Specifications

Specifications

HPV6 L1
ELISA -1:5000-1:10000
Unconjugated
Rabbit
Virus
Recombinant Human Papilloma Virus type 16 L1 virus like particle (HPV16 L1 VLP)
Monoclonal
Protein A purified
1
ELISA
Purified
RUO
PBS, pH7.0
IgG
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit