Learn More
Advanced ImmunoChemical, Inc. Human Papillomavirus L1 Protein (HPVL1), Recombinant, Type 16, Source:Purity > 90 %, 0.1mg
Supplier:  Advanced ImmunoChemical, Inc. 8HPVL160.1MG
Encompass_Preferred
This item is manufactured by HyTest, Ltd and distributed by Advanced ImmunoChemical, Inc. Antigens / Infectious Diseases / Human Papillomavirus L1 protein (HPVL1), recombinant, type 16. Source: Purity > 90 % (SDS-PAGE).Each lot is tested for protein content (Lowry method), lipid content (Total lipid), purity (SDS-PAGE), carbohydrate content (Anthrone method), antigen content (Immunoblot). Immunological activity: Tested in quantitative immunoblot with anti-HPVL1 antibodies. Purification: Purification is conducted from broken yeast cells by following scheme: clarification, ammonium sulfate precipitation, ultrafiltration, cation exchange chromatography, zonal centrifugation, size- exclusion chromatography, sterile filtration. Presentation: 20 mM KH2PO4 (pH 7.2), 320 mM NaCl, 1.7 mM EDTA, 0.01% Tween-80, 10 mM histidine. Storage Conditions: + 4 °C (between +2 and +8 °C allowed)
Catalog No. 50-153-9827