Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HR23A/Rad23A TUBE1 Antibody (SR1997), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $413.50
Specifications
Antigen | HR23A/Rad23A TUBE1 |
---|---|
Clone | SR1997 |
Dilution | Western Blot 1:1000-1:5000, Flow Cytometry 1:50 |
Applications | Western Blot, Flow Cytometry |
Classification | Monoclonal |
Description
HR23A/Rad23A TUBE1 Monoclonal antibody specifically detects HR23A/Rad23A TUBE1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow CytometrySpecifications
HR23A/Rad23A TUBE1 | |
Western Blot 1:1000-1:5000, Flow Cytometry 1:50 | |
Monoclonal | |
Purified | |
RUO | |
PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
5886 | |
IgG | |
Affinity purified |
SR1997 | |
Western Blot, Flow Cytometry | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
hHR23A, HR23A | |
A synthesized peptide derived from human HR23A/Rad23A TUBE1 (Uniprot #: P54725) | |
Primary | |
Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title