Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HR6A/UBE2A Rabbit anti-Human, Janelia Fluor 549, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP300042JF549

Catalog No. NB064080


Add to cart

Description

Description

HR6A/UBE2A Polyclonal antibody specifically detects HR6A/UBE2A in Human samples. It is validated for Immunofluorescence
Specifications

Specifications

HR6A/UBE2A
Janelia Fluor 549
50mM Sodium Borate
EC 6.3.2.19, hHR6A, HR6A, RAD6ARAD6 homolog A, UBC2, Ubiquitin carrier protein A, ubiquitin-conjugating enzyme E2 A, ubiquitin-conjugating enzyme E2A (RAD6 homolog), Ubiquitin-protein ligase A
Produced in rabbits immunized with purified, recombinant HR6A/UBE2A (P49459; Met1-Cys152).
Affinity purified
RUO
Store at 4°C in the dark.
Polyclonal
Human
Immunofluorescence
Immunocytochemistry/Immunofluorescence
Purified
Rabbit
IgG
0.1 mL
DNA Repair, Ubiquitin Proteasome Pathway
Primary
7319
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit