Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ HR6A/UBE2A Recombinant Protein Antigen
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Supplier: Novus Biologicals™ NBP257290PEP
Description
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human HR6A/UBE2A. Source: E.coli Amino Acid Sequence: APSENNIMVWNAVIFGPEGTPFEDGTFK The HR6A/UBE2A Recombinant Protein Antigen is derived from E. coli. The HR6A/UBE2A Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.Specifications
7319 | |
HR6A/UBE2A Recombinant Protein Antigen | |
PBS and 1M Urea, pH 7.4. | |
EC 6.3.2.19, hHR6A, HR6A, RAD6ARAD6 homolog A, UBC2, Ubiquitin carrier protein A, ubiquitin-conjugating enzyme E2 A, ubiquitin-conjugating enzyme E2A (RAD6 homolog), Ubiquitin-protein ligase A | |
Unlabeled | |
100μL | |
E.coli |
>80% by SDS-PAGE and Coomassie blue staining | |
Store at −20°C. Avoid freeze-thaw cycles. | |
Blocking/Neutralizing, Control | |
UBE2A | |
Recombinant Protein Antigen | |
RUO | |
This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-50989. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?
Provide Content Correction