Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HR6B/UBE2B Mouse anti-Human, Alexa Fluor™ 647, Clone: OTI1E11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271337AF647

Catalog No. N271337A647


Add to cart

Description

Description

HR6B/UBE2B Monoclonal antibody specifically detects HR6B/UBE2B in Human samples. It is validated for Western Blot.
Specifications

Specifications

HR6B/UBE2B
OTI1E11
Western Blot
Affinity Purified, Conjugated
UBE2B
Full length recombinant protein of human HR6B/UBE2B (NP_003328) produced in E.coli.
Immunogen affinity purified
Biologically Active Proteins
Primary
7320.0
Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
E2 protein, E2-17kDa, HHR6B, HR6B, RAD6 homolog B, RAD6BEC 6.3.2.19, UBC2, Ubiquitin carrier protein B, ubiquitin-conjugating enzyme E2 B, Ubiquitin-conjugating enzyme E2-17 kDa, ubiquitin-conjugating enzyme E2B (RAD6 homolog), Ubiquitin-protein ligase B
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit