Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HRAS Rabbit anti-Human, Alexa Fluor 647, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP299921AF647

Catalog No. NB063533


Add to cart

Description

Description

HRAS Polyclonal antibody specifically detects HRAS in Human samples. It is validated for Western Blot, ELISA
Specifications

Specifications

HRAS
Alexa Fluor 647
50mM Sodium Borate
Rabbit
IgG
0.1 mL
Cell Cycle and Replication, Metastasis, mTOR Pathway
Primary
3265
ELISA, Western Blot
Western Blot, ELISA
Purified
Produced in rabbits immunized with purified, recombinant HRAS (P01112; Met1-Cys186).
Affinity purified
RUO
Store at 4°C in the dark.
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit