Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HRAS Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen HRAS
Applications Immunofluorescence, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB093300
View Documents
Novus Biologicals
NBP304486100UL
100 μg
Each for $320.71
Add to cart
 
NB093301
View Documents
Novus Biologicals
NBP30448620UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

HRAS Polyclonal antibody specifically detects HRAS in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications

Specifications

HRAS
Unconjugated
Rabbit
Cell Cycle and Replication, Metastasis, mTOR Pathway
PBS with 50% glycerol, pH7.3.
Recombinant fusion protein containing a sequence corresponding to amino acids 1-189 of human HRAS (NP_005334.1).
Polyclonal
Affinity purified
Immunofluorescence, Western Blot
Purified
RUO
Human, Mouse, Rat
3265
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit