Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HRASLS Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562339
Description
Immunogen sequence: DGYVINIAPV DGIPASFTSA KSVFSSKALV KMQLLKDVVG NDTYRINNKY DETYPPLPVE EIIKRSEFVI GQEVAYNLLV NN Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.

Specifications
HRASLS | |
Polyclonal | |
Unconjugated | |
HRASLS | |
A-C1; HRAS like suppressor; HRAS-like suppressor 1; HRASLS; HRASLS1; H-REV107; H-REV107 protein-related protein; HRSL1; HSD28; phospholipase A; Phospholipase A and acyltransferase 1; Phospholipid-metabolizing enzyme A-C1; PLAAT1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57110 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HDD0 | |
HRASLS | |
Recombinant protein corresponding to Human HRASLS. Recombinant protein control fragment (Product #RP-101248). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction