Learn More
HRASLS purified MaxPab mouse Mouse anti-Human, Polyclonal, Abnova™
Mouse Polyclonal Antibody
Manufacturer:  Abnova Corporation H00057110B01P
Catalog No. 89-032-164
Add to cart
Description
Description
Mouse polyclonal antibody raised against a full-length human HRASLS protein.
  • High specificity
  • High reactivity
Specifications
Specifications
HRASLS purified MaxPab mouse
Unconjugated
In 1x PBS, pH 7.4
Liquid
HRASLS
HRASLS (NP_065119.1, 1 a.a. ~ 168 a.a) full-length human protein.
RUO
Primary
57110
Western Blot
Mouse polyclonal antibody raised against a full-length human HRASLS protein.
NM_020386.2
A-C1|H-REV107|HRASLS1|HSD28
Mouse
50 ug
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)