Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HRASLS3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567562
Description
HRASLS3 Polyclonal Antibody for IHC (P)
HRASLS3 specifically catalyzes the release of fatty acids from phospholipids in adipose tissue. It also has a weak lysophospholipase activity. Tumor suppressor that may be involved in interferon-dependent cell death.
Specifications
HRASLS3 | |
Polyclonal | |
Unconjugated | |
PLAAT3 | |
Adipose-specific phospholipase A2; adipose-specific PLA2; AdPLA; C78643; Ca-independent phospholipase A1/2; group XVI phospholipase A1/A2; Group XVI phospholipase A2; HRAS like suppressor 3; HRAS-like suppressor 1; HRAS-like suppressor 3; HRASLS3; H-rev 107 protein; H-rev 107 protein homolog; Hrev107; H-REV107; H-REV107-1; HREV107-1; HREV107-3; HRSL3; LRAT-like protein 3; MLP-3; Phospholipase A and acyltransferase 3; phospholipase A2 group XVI; phospholipase A2, group XVI; Pla2g16; PLAAT3; Renal carcinoma antigen NY-REN-65 | |
Rabbit | |
affinity chromatography | |
RUO | |
11145 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.5 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P53816 | |
PLAAT3 | |
Synthesized peptide derived from human HRASLS3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction