Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HRC Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548433

Catalog No. PA548433

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May play a role in the regulation of calcium sequestration or release in the SR of skeletal and cardiac muscle.
Specifications

Specifications

HRC
Polyclonal
PBS with 0.09% sodium azide
P23327
Sarcoplasmic reticulum histidine-rich calcium-binding protein, HRC, HCP
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3270
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
HRC
Liquid
HRC
KLH conjugated synthetic peptide between 277-306 amino acids from the Central region of human HRC.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit