Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

HRC Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 181421AP

Catalog No. 50-173-2010



Description

Description

May play a role in the regulation of calcium sequestration or release in the SR of skeletal and cardiac muscle.
Specifications

Specifications

HRC
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P23327
HCP, HRC
Rabbit
IgG
150 μL
-20°C
Primary
15464, 3270
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
HRC
Liquid
Hrc
HRC Fusion Protein Ag12794
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit