Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HRH2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533844
Description
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Orangutan, Gibbon, Monkey (100%) Mouse, Rat, Hamster (94%).
H2 Histamine Receptor stimulates adenylate cyclase activity and leads to the release of acids in the stomach. It is also involved in GI motility and intestinal secretion. Treatments for acid-peptic disorders are based on the inhibition of histamine H2 receptor activity. The 3-untranslated region may be involved in differential expression. Histamine H2 receptor has been reported to be expressed in bone marrow, brain, heart, lymph node, nose, placenta, spinal cord, and stomach. An EST has been isolated from a kidney library.
Specifications
HRH2 | |
Polyclonal | |
Unconjugated | |
HRH2 | |
gastric receptor 1; gastric receptor I; H2R; HH2R; histamine H2 receptor; histamine receptor; histamine receptor H 2; histamine receptor H2; HRH2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3274 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P25021 | |
HRH2 | |
Synthetic 16 amino acid peptide from 3rd cytoplasmic domain of human HRH2 / Histamine H2 Receptor. | |
50 μg | |
Primary | |
Human, Monkey | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction