Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HRP-1/HDGFL1 Rat anti-Mouse, Alexa Fluor™ 350, Clone: RM0092-5B24, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP212354AF350

Catalog No. NP21234A30


Add to cart

Description

Description

HRP-1/HDGFL1 Monoclonal antibody specifically detects HRP-1/HDGFL1 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

HRP-1/HDGFL1
RM0092-5B24
Western Blot
Conjugated, Purified
HDGFL1
Mouse HDGFL1
Protein G purified
Store at 4°C in the dark.
Monoclonal
Mouse
Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
dJ309H15.1, HDGF (hepatoma-derived growth factor) like, hepatoma derived growth factor-like 1, hepatoma-derived growth factor-like protein 1, PWWP domain containing 1, PWWP domain-containing protein 1, PWWP1
Rat
IgG2
0.1 mL
Primary
154150.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit