Learn More
HRP-1/HDGFL1 Rat anti-Mouse, Alexa Fluor™ 594, Clone: RM0092-5B24, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212354AF594
Catalog No. NP21234A94
Add to cart
Description
Description
HRP-1/HDGFL1 Monoclonal antibody specifically detects HRP-1/HDGFL1 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
HRP-1/HDGFL1
RM0092-5B24
Western Blot
dJ309H15.1, HDGF (hepatoma-derived growth factor) like, hepatoma derived growth factor-like 1, hepatoma-derived growth factor-like protein 1, PWWP domain containing 1, PWWP domain-containing protein 1, PWWP1
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Mouse
Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
HDGFL1
Mouse HDGFL1
0.1 mL
Primary
154150