Learn More
Seracare Life Sciences IncSupplier Diversity Partner HRP ELISA KIT ANTI-MOUSE  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 51100011
Protein Detector ELISA Kits offer a convenient starting point for the development of microwell ELISA protocols for antigen and antibody detection. These kits provide pretested components and suggested protocols to facilitate the development of ELISA procedures. KPLs protein Detector ELISA Kit, HRP Systems are provided with ABTS Peroxidase Substrate which offers performance, reliability, and low background. Highly sensitive reagents optimized for antigen detection in picogram quantities.
Catalog No. 50-675-13