Learn More
Seracare Life Sciences IncSupplier Diversity Partner HRP-LABELED STREPTAVIDIN  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 55500001
Streptavidin labeled with horseradish peroxidase. Streptavidin is a protein isolated from the bacterium Streptomyces avidinii and has no carbohydrate moieties. It has a molecular weight of approximately 60,000 Daltons with an isoelectric point of 5. Product is in liquid form at use dilution and is specifically formulated for immunohistochemistry applications.
Catalog No. 50-675-75