Learn More
Seracare Life Sciences IncSupplier Diversity Partner HRP-RABBIT ANTI-VIBRIO SPECIES  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 53200028
Protein G purified BacTrace antibody to Vibrio species labeled with Horseradish Peroxidase (HRP). Isolated from a serum pool of rabbits immunized with heat-killed whole cells of various strains of Vibrio.
Catalog No. 50-669-89