Learn More
Leap PAL PartsSupplier Diversity Partner LMARK Headspace Crimp Cap 20mm PTFE/Clear Silicone, 3mm, 100 Pack  DFS Item
Manufacturer:  Leap PAL PartsSupplier Diversity Partner XAR20039982
Cap, Crimp, Headspace, Standard, 0.005 in. PTFE/Clear Silicone - 3.2mm, 100 Pack
Catalog No. 50-635-123