Learn More
Leap PAL PartsSupplier Diversity Partner Headspace Pressure relief Crimp Cap 20mm Blue Silicone/White PTFE, 3mm, 100 Pack  DFS Item
Manufacturer:  Leap PAL PartsSupplier Diversity Partner XAR20038622
L-MARK Brand Cap Crimp 20mm 3.0mm Blue Silicone/White PTFE Pressure relief100 Pack
Catalog No. 50-635-109