Learn More
Leap PAL PartsSupplier Diversity Partner Headspace Magnetic Crimp Cap 20mm, 5mm hole, PTFE/Gray Butyl, 3mm, 100 Pack  DFS Item
Manufacturer:  Leap PAL PartsSupplier Diversity Partner XAR20038559
L-MARK Brand Cap Crimp 20mm 3.0mm PTFE/Gray Butyl Magnetic With 5mm hole100 Pack
Catalog No. 50-635-131