Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HS6ST2 Polyclonal Antibody
HS6ST2 Polyclonal Antibody
Supplier: Thermo Scientific PA547372
Description
HS6ST2 Polyclonal Antibody for Western Blot, IP

Specifications
HS6ST2 | |
Polyclonal | |
Unconjugated | |
HS6ST2 | |
6OST2; EC-1; heparan sulfate 6-O-sulfotransferase 2; heparan-sulfate 6-O-sulfotransferase 2; Hs6st2; HS6ST-2; LOW QUALITY PROTEIN: heparan-sulfate 6-O-sulfotransferase 2; PSEC0092; RGD1563872; RGD1564397 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
90161 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q96MM7 | |
HS6ST2 | |
Mouse myeloma cell line NS0-derived recombinant human Heparan Sulfate 6-O-Sulfotransferase 2/HS6ST2 Gln179-Arg605. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction