Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Hs6st3 Polyclonal Antibody
Sheep Polyclonal Antibody
Supplier: Invitrogen™ PA547733
Description
Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
6-O-sulfation enzyme which catalyzes the transfer of sulfate from 3'-phosphoadenosine 5'-phosphosulfate (PAPS) to position 6 of the N-sulfoglucosamine residue (GlcNS) of heparan sulfate.
Specifications
Hs6st3 | |
Polyclonal | |
Unconjugated | |
Hs6st3 | |
6OST3; 6-ost-3; DKFZp761K2315; heparan sulfate 6-O-sulfotransferase 3; heparan sulfate 6-O-sulfotransferase 3b; heparan-sulfate 6-O-sulfotransferase 3; heparan-sulfate 6-O-sulfotransferase 3 pseudogene; heparan-sulfate 6-O-sulfotransferase 3-B; HS 6-OST-3-B; hs6st3; HS6ST-3; hs6st3b; RGD1560050; sb:eu962 | |
Sheep | |
Antigen Affinity Chromatography | |
RUO | |
50787 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Frozen) | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9QYK4 | |
Hs6st3 | |
CHO-derived recombinant mouse Heparan Sulfate 6-O-Sulfotransferase/HS6ST3 Pro28-Trp470. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction