Learn More
Abnova™ HSCB 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00150274T01
Catalog No. 89-947-925
Add to cart
Specifications
Specifications
293T
NM_172002
pCMV-HSCB full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
HSCB 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 27.4kDa
Transient overexpression cell lysate was tested with anti-HSCB antibody (Mfr. No. H00150274-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human